Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII) (PubMed:20855544, PubMed:21768307). May act at an RNAP assembly step prior to nuclear import (PubMed:21768307). Forms an interface between the RNA polymerase II enzyme and chaperone/scaffolding proteins, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation (PubMed:17643375). May be involved in nuclear localization of XPA (PubMed:11058119).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP(GTP)-binding protein; GPN-loop GTPase 1; MBD2-interacting protein; MBDin; methyl-CpG binding domain protein 2 interacting protein; RNA polymerase II associated protein 4; RNAPII-associated protein 4; XPA binding protein 1; XPA binding protein 1, GTPase; XPA-binding protein 1
Gene Aliases: 2410004J02Rik; AI449615; ATPBD1A; GPN1; HUSSY-23; MBDIN; NTPBP; RPAP4; XAB1
UniProt ID: (Human) Q9HCN4, (Mouse) Q8VCE2
Entrez Gene ID: (Human) 11321, (Mouse) 74254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support