Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Elephant, Panda, Horse, Pig (100%) Bovine (94%).
GPR137 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor TM7SF1L1; Integral membrane protein GPR137; Transmembrane 7 superfamily member 1-like 1 protein
Gene Aliases: AI428855; C11orf4; GPR137; GPR137A; TM7SF1L1
UniProt ID: (Human) Q96N19, (Mouse) Q80ZU9
Entrez Gene ID: (Human) 56834, (Pig) 100524971, (Horse) 100050142, (Dog) 612124, (Mouse) 107173, (Rat) 689984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support