Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Urotensin-II receptor has been reported to be expressed at high levels in the heart and pancreas and also in artery and smooth muscle. Low transcript levels have been identified in brain, particularly in thalamus, and superior occipital gyrus. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 14; G-protein coupled receptor 14; G-protein coupled sensory epithelial neuropeptide-like receptor; putative G protein-coupled receptor (SENR); SENR; Uii-r; Uii-r1a; UII-receptor; Uiir; UR-2-R; UR-II-R; UR2R; Urotensin II receptor; Urotensin-2 receptor; UT-II; UTIIR
Gene Aliases: GPR14; Senr; UR-2-R; UTR; UTR2; UTS2R
UniProt ID: (Human) Q9UKP6, (Rat) P49684, (Mouse) Q8VIH9
Entrez Gene ID: (Human) 2837, (Rat) 57305, (Mouse) 217369
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support