Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTQAGRRGPG TPEPRPRTQP MASPRLGTFC CPTRDAATQL VLSFQPRAFH ALCLGSGGLR LALGLLQLLP GRRPAGPGSP ATSPPASVRI LRAAAACDLL GCLGMVIRST VWLGFPNFVD SVSDMNHTEI WPAAFCVGSA MWIQLLYSAC FWWLFCYAVD AYLVIRRSAG LSTILLYHIM AWGLATLLCV EGAAMLYYPS VSRCERGLDH AIPHYVTMYL PLLLVLVANP ILFQKTVTAV ASLLKGRQGI YTENERRMGA VIKIRFFKIM LVLIICWLSN IINESLLFYL EMQTDINGGS LKPVRTAAKT TWFIMGILNP AQGFLLSLAF YGWTGCSLGF QSPRKEIQWE SLTTSAAEGA HPSPLMPHEN PASGKVSQVG GQTSDEALSM LSEGSDASTI EIHTASESCN KNEGDPALPT HGDL
Ocular albinism type 1 protein is a conserved integral membrane protein with seven transmembrane domains. It is expressed in the eye and epidermal melanocytes. Defects in GPR143 are the cause of ocular albinism type 1 (OA1); also known as Nettleship-Falls type ocular albinism. OA1 is an X-linked disorder characterized by severe impairment of visual acuity, retinal hypopigmentation and the presence of macromelanosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor 143; ocular albinism 1; Ocular albinism type 1 protein
Gene Aliases: GPR143; NYS6; OA1
UniProt ID: (Human) P51810
Entrez Gene ID: (Human) 4935
Molecular Function: G-protein coupled receptor transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support