Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
GPR171 is a protein coding gene. Orphan receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F730001G15Rik; G-protein coupled receptor 171; G-protein coupled receptor H963; platelet activating receptor homolog; probable G-protein coupled receptor 171
Gene Aliases: A630006E05; F730001G15Rik; GPR171; H963
UniProt ID: (Human) O14626, (Mouse) Q8BG55
Entrez Gene ID: (Human) 29909, (Rat) 688737, (Mouse) 229323
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support