Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LIAPGPTTA VSYMSVKCVD ARKNHHKTKW FVPWGPNHCD KIRDIEEAIP REIEANDIVF SVHIPLPHME MSPWFQFMLF ILQLDIAFKL NNQIRENAEV SMDVSLAYRD DAFAEWTEMA HERVPRKLKC TFTSPKTPEH EGRYYECDVL PFMEIGSVAH KFYLLNIRLP VNEKKKINVG IGEIKDIRLV GIHQNGGFTK VWFAMKT (34-239 aa encoded by BC110826)
Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: evenness interrupted homolog; EVI; G protein-coupled receptor 177; Integral membrane protein GPR177; Protein evenness interrupted homolog; Protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373
Gene Aliases: 5031439A09Rik; AI173978; AI987742; C1orf139; EVI; GPR177; mig-14; MRP; UNQ85/PRO18667; WLS
UniProt ID: (Human) Q5T9L3, (Mouse) Q6DID7, (Rat) Q6P689
Entrez Gene ID: (Human) 79971, (Mouse) 68151, (Rat) 362065
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support