Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total GPR45.
This intronless gene encodes a member of the G protein-coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins. This protein may function in the central nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: high-affinity lysophosphatidic acid receptor; Probable G-protein coupled receptor 45; PSP24-1; PSP24-alpha; PSP24alpha
Gene Aliases: 9230112G11Rik; AW046772; GPR45; PSP24; PSP24(ALPHA); PSP24A
UniProt ID: (Human) Q9Y5Y3, (Mouse) Q9EQQ4
Entrez Gene ID: (Human) 11250, (Mouse) 93690
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support