Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GPR56 is a member of the G protein-coupled receptor family. The protein contains 7 transmembrane domains and a mucin-like domain in the N-terminal region. The gene is implicated in the regulation of brain cortical patterning. The protein binds specifically to transglutaminase 2 in the extracellular space. Expression of this gene is downregulated in melanoma cell lines, and overexpression of this gene can suppress tumor growth and metastasis. Mutations in this gene result in bilateral frontoparietal polymicrogyria. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7-transmembrane protein with no EGF-like N-terminal domains-1; Adhesion G-protein coupled receptor G1; Cyt28; DKFZp781L1398; G protein-coupled receptor 56; G-protein coupled receptor 56; Protein TM7XN1; testicular tissue protein Li 77
Gene Aliases: ADGRG1; BFPP; BPPR; GPR56; TM7LN4; TM7XN1; UNQ540/PRO1083
UniProt ID: (Human) Q9Y653
Entrez Gene ID: (Human) 9289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support