Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals. Has a higher affinity for neuropeptide B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 7; G-protein coupled receptor 7; MGC129755; neuropeptide B/W receptor 1; Neuropeptides B/W receptor type 1; opioid-somatostatin-like receptor 7
Gene Aliases: GPR7; Nbpwr1; NPBWR1
UniProt ID: (Human) P48145, (Mouse) P49681, (Rat) Q56UD9
Entrez Gene ID: (Human) 2831, (Mouse) 226304, (Rat) 297795
Molecular Function: G-protein coupled receptor transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support