Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSKVIRTNN ALYFAFHWFA MSSTCYNPFI YCWLNENFRI ELKALLSMCQ RPPKPQEDRP PSPVPSFRVA WTEKNDGQRA PLANNLLPTS QLQSGKTDLS SVEPIVTMS (316-423 aa encoded by BC067473)
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 72; G-protein coupled receptor 72; G-protein coupled receptor 83; glucocorticoid induced receptor; Hcepto9; Jp05
Gene Aliases: GIR; GPR72; GPR83; JP05; KIAA1540
UniProt ID: (Human) F5GZ43
Entrez Gene ID: (Human) 10888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support