Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
PA1-32840 detects GPCR SH120 from human samples.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 89; G protein-coupled receptor 89B; G protein-coupled receptor 89C; Golgi pH regulator; Golgi pH regulator A; Golgi pH regulator B; Protein GPR89; Protein GPR89A; Protein GPR89B; putative Golgi pH regulator C; Putative MAPK-activating protein PM01; Putative NF-kappa-B-activating protein 90; putative protein GPR89C
Gene Aliases: CGI-13; GPHR; GPHRA; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; RCJMB04_3g11; RGD1304837; SH120; UNQ192; UNQ192/PRO218
UniProt ID: (Chicken) Q5F448, (Human) B7ZAQ6
Entrez Gene ID: (Chicken) 418459, (Human) 51463, (Rat) 362003
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support