Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: GIPADPNNV FLSTGASDAI VTVLKLLVAG EGHTRTGVLI PIPQYPLYSA TLAELGAVQV DYYLDEERAW ALDVAELHRA LGQARDHCRP RALCVINPGN PTGQVQTREC IEAVIRFAFE ERLFLLADEV YQDNVYAAGS QFHSFKKVLM EMGPPYAGQQ ELASFHSTSK GYMGECGFRG GYVEVVNMDA AVQQQMLKLM SVRLCPPVPG QALLDLVVSP PAPTDPSFAQ FQAEKQAVLA ELAAKAKLTE QVFNEAPGIS CNPVQGAMYS FPRVQLPPRA VERAQELGLA PDMFFCLRLL EETGICVVPG SGFGQREGTY HFRMTILPPL EKLRLLLEKL SRFHAKFTLE YS (146-496 aa encoded by BC018207)
GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alanine aminotransferase; Alanine aminotransferase 1; ALT1; Glutamate pyruvate transaminase 1; glutamic pyruvate transaminase; glutamic pyruvic transaminase (alanine aminotrasferase); glutamic pyruvic transaminase 1, soluble; Glutamic--alanine transaminase 1; Glutamic--pyruvic transaminase 1; glutamic-alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); GPT 1
Gene Aliases: 1300007J06Rik; 2310022B03Rik; AAT1; ALT; ALT1; GPT; Gpt-1; GPT1
UniProt ID: (Human) P24298, (Rat) P25409, (Mouse) Q8QZR5
Entrez Gene ID: (Human) 2875, (Rat) 81670, (Mouse) 76282
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support