Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: HepG2.
This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastrointestinal glutathione peroxidase; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); Glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP-2; GPx-2; GPx-GI; HGNC:4554; intestinal GPx; Phospholipid hydroperoxide glutathione peroxidase GPX2
Gene Aliases: GI-GPx; GPRP; GPRP-2; GPx-2; GPx-GI; GPX2; GSHPx-2; GSHPX-GI
UniProt ID: (Human) P18283, (Mouse) Q9JHC0
Entrez Gene ID: (Human) 2877, (Mouse) 14776
Molecular Function: peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support