Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa whole cell extract.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CL683; FLJ14777; glutathione peroxidase 6; Glutathione peroxidase 7; GPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase
Gene Aliases: CL683; GPx-7; GPX6; GPX7; GSHPx-7; NPGPx; UNQ469/PRO828
UniProt ID: (Human) Q96SL4
Entrez Gene ID: (Human) 2882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support