Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: RGINEQGLYR IVGVNSRVQK LLSVLMDPKT ASETETDICA EWEIKTITSA LKTYLRMLPG PLMMYQFQRS FIKAAKLENQ ESRVSEIHSL VHRLPEKNRQ MLQLLMNHLA NVANNHKQNL MTVANLGVVF GPTLLRPQEE TVAAIMDIKF QNIVIEILIE NHEKIFNTVP DMPLTNAQLH LSRKKSSDSK PPSCSERPLT LFHTVQSTEK QEQRNSIINS SLESVSSNPN SILNSSSSLQ PNMNSSDPDL AVVKPTRPNS LPPNPSPTSP LSPSWPMFSA PSSPMPTSST SSDSSPVSTP FRKAKALYAC KAEHDSELSF TAGTVFDNVH PSQEPGWLEG TLNGKTGLIP ENYVEFL
Interaction of a cell with the extracellular matrix triggers integrin cell surface receptors to begin signaling cascades that regulate the organization of the actin-cytoskeleton. One of the proteins involved in these cascades is focal adhesion kinase. The protein encoded by this gene is a GTPase activating protein that binds to focal adhesion kinase and mediates the activity of the GTP binding proteins RhoA and Cdc42. Defects in this gene are a cause of juvenile myelomonocytic leukemia (JMML). Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ42530; GRAF1; GTPase regulator associated with focal adhesion kinase; GTPase regulator associated with focal adhesion kinase pp125(FAK); Oligophrenin-1-like protein; Rho GTPase-activating protein 26; Rho-type GTPase-activating protein 26
Gene Aliases: ARHGAP26; GRAF; GRAF1; KIAA0621; OPHN1L; OPHN1L1
UniProt ID: (Human) Q9UNA1
Entrez Gene ID: (Human) 23092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support