Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • GRB2 Antibodies

            Proteintech

            GRB2 Polyclonal Antibody

            17 References
            View all (21) GRB2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite GRB2 Polyclonal Antibody

            • Antibody Testing Data (8)
            Application
            GRB2 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 8

            GRB2 Antibody (10254-2-AP) in ICC/IF

            Immunofluorescent analysis of HEK-293 cells using 10254-2-AP (GRB2 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            GRB2 Antibody in Immunocytochemistry (ICC/IF)
            GRB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            GRB2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            GRB2 Antibody in Western Blot (WB)
            GRB2 Antibody in Western Blot (WB)
            GRB2 Antibody in Western Blot (WB)
            GRB2 Antibody in Western Blot (WB)
            GRB2 Antibody in Western Blot (WB)

            Product Details

            10254-2-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 10 publications 10 publications

            Immunohistochemistry (IHC)

            -
            View 4 publications 4 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 1 publication 1 publication

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            GRB2 Fusion Protein Ag0395 (1-214 aa encoded by BC000631)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.33 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MEAIAKYDF KATADDELSF KRGDILKVLN EECDQNWYKA ELNGKDGFIP KNYIEMKPHP WFFGKIPRAK AEEMLSKQRH DGAFLIRESE SAPGDFSLSV KFGNDVQHFK VLRDGAGKYF LWVVKFNSLN ELVDYHRSTS VSRNQQIFLR DIEQVPQQPT YVQALFDFDP QEDGELGFRR GDFIHVMDNS DPNWWKGACH GQTGMFPRNY VTPVN (1-214 aa encoded by BC000631)

            Target Information

            GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: abundant SRC homology; Adapter protein GRB2; ASH protein; epidermal growth factor receptor-binding protein GRB2; Growth factor receptor-bound protein 2; growth factor receptor-bound protein 3; HT027; OTTHUMP00000166097; OTTHUMP00000166098; Protein Ash; SH2/SH3 adapter GRB2

            View more View less

            Gene Aliases: AA408164; ASH; EGFRBP-GRB2; GRB2; Grb3-3; MST084; MSTP084; NCKAP2

            View more View less

            UniProt ID: (Human) P62993, (Mouse) Q60631, (Rat) P62994

            View more View less

            Entrez Gene ID: (Human) 2885, (Mouse) 14784, (Rat) 81504

            View more View less

            Function(s)

            SH3/SH2 adaptor activity Ras guanyl-nucleotide exchange factor activity epidermal growth factor receptor binding neurotrophin TRKA receptor binding protein binding 1-phosphatidylinositol-3-kinase activity SH3 domain binding protein kinase binding protein phosphatase binding identical protein binding insulin receptor substrate binding poly(A) RNA binding ephrin receptor binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity phosphotyrosine binding protein domain specific binding phosphoprotein binding scaffold/adaptor protein

            Process(es)

            MAPK cascade epidermal growth factor receptor signaling pathway Ras protein signal transduction cell-cell signaling axon guidance insulin receptor signaling pathway fibroblast growth factor receptor signaling pathway positive regulation of signal transduction regulation of phosphatidylinositol 3-kinase signaling viral process positive regulation of actin filament polymerization T cell costimulation receptor internalization phosphatidylinositol-3-phosphate biosynthetic process Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis ERBB2 signaling pathway negative regulation of epidermal growth factor receptor signaling pathway signal transduction in response to DNA damage regulation of MAPK cascade positive regulation of GTPase activity phosphatidylinositol phosphorylation phosphatidylinositol-mediated signaling anatomical structure formation involved in morphogenesis leukocyte migration branching involved in labyrinthine layer morphogenesis cellular response to ionizing radiation positive regulation of reactive oxygen species metabolic process aging cell differentiation protein heterooligomerization
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0