Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
RAS (MIM 190020) GTPases cycle between an inactive GDP-bound state and an active GTP-bound state. Guanine-nucleotide exchange factors (GEFs), such as RASGRFs, stimulate the conversion of the GDP-bound form into the active form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanine nucleotide-releasing factor 2; Ras guanine nucleotide exchange factor 2; Ras protein-specific guanine nucleotide-releasing factor 2; Ras-GRF2; Ras-specific guanine nucleotide-releasing factor 2
Gene Aliases: 6330417G04Rik; AW048350; GRF2; RAS-GRF2; RASGRF2
UniProt ID: (Human) O14827, (Mouse) P70392, (Rat) Q99JE4
Entrez Gene ID: (Human) 5924, (Mouse) 19418, (Rat) 114513
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support