Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: GRID1.
GRID1 is a subunit of glutamate receptor channels. These channels mediate most of the fast excitatory synaptic transmission in the central nervous system and play key roles in synaptic plasticity. GRID1 is a receptor for glutamate. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. The postsynaptic actions of Glu are mediated by a variety of receptors that are named according to their selective agonists.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluD1; gluR delta-1 subunit; glutamate receptor; glutamate receptor delta-1 subunit; Glutamate receptor ionotropic, delta-1; glutamate receptor, ionotropic, delta 1
Gene Aliases: GluD1; GluRdelta1; GRID1; KIAA1220
UniProt ID: (Human) Q9ULK0, (Mouse) Q61627, (Rat) Q62640
Entrez Gene ID: (Human) 2894, (Mouse) 14803, (Rat) 79219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support