Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
MA5-31746 has been tested in indirect ELISA.
This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAA1; Excitatory amino acid receptor 1; GluK4; glutamate receptor; Glutamate receptor ionotropic, kainate 4; Glutamate receptor KA-1; glutamate receptor KA1; glutamate receptor, ionotropic, kainate 4; KA1
Gene Aliases: EAA1; GluK4; GRIK; GRIK4; KA1
UniProt ID: (Human) Q16099
Entrez Gene ID: (Human) 2900
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support