Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PFRQHKTKD KHEIDRMTLT MAVELPDSFS PELRSLLEGL LQRDVNRRLG CLGRGAQEVK ESPFFRSLDW QMVFLQKYPP PLIPPRGEVN AADAFDIGSF DEEDTKGIKL LDSDQELYRN FPLTISERWQ QEVAETVFDT INAETDRLEA RKKAKNKQLG HEEDYALGKD CIMHGYMSKM GNPFLTQWQR RYFYLFPNRL EWRGEGEAPQ SLLTMEEIQS VEETQIKERK CLLLKIRGGK QFILQCDSDP ELVQWKKELR DAYREAQQLV QRVPKMKNKP RSPVVELSKV PLVQRGSANG L (390-689 aa encoded by BC037963)
ADRBK1 (GRK2) is a serine/threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs). G protein-coupled receptor kinases (GRKs) are important regulators of G protein-coupled receptors(GPCRs). GRK2 (83 kDa), one of six members of this family that have been identified, is ubiquitously expressed in mammals. After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs (e.g., a2-adrenergic receptor) are phosphorylated by GRKs. Internalization of the GPCRs, regulated by beta-arrestin-1, leads to activation of the Ras - Raf - ERK1&2 signaling pathway. GRK2 activity is tightly controlled by different mechanisms including phosphorylation by kinases such as PKC, Src and ERK1&2, as well as interaction with various proteins. ERK phosphorylates and thus inactivates GRK2 on serine 670 in a negative feedback mechanism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic beta receptor kinase 1; adrenergic, beta, receptor kinase 1; Beta-adrenergic receptor kinase 1; Beta-ARK-1; EC 2.7.1.126; G-protein coupled receptor kinase 2; grk 2
Gene Aliases: ADRBK1; BARK; BARK1; BETA-ARK1; GRK2
UniProt ID: (Human) P25098
Entrez Gene ID: (Human) 156
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support