Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
MA1-2013 detects the beta andrenergic receptor kinase 1 in human, mouse and rat samples.
MA1-2013 has been successfully used in Western blot, immunocytochemistry, immunofluoresence, immunoprecipitation, and ELISA procedures. By Western blot this antibody detects a ~30 kDa protein representing the beta andrenergeic receptor kinase 1 in human HeLa cells. In immunocytochemistry procedures MA1-2013 recognizes the beta andrenergic receptor kinase 1 in HeLa, Hek-293 and 3T3 cells. Immunofluoresence in HeLa cells yeilds predominantely cytoplasmic membrane associated staining with some staining in the ruffles.
The MA1-2013 immunogen is recombinant beta adrenergic receptor kinase 1.
ADRBK1 (GRK2) is a serine/threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs). G protein-coupled receptor kinases (GRKs) are important regulators of G protein-coupled receptors(GPCRs). GRK2 (83 kDa), one of six members of this family that have been identified, is ubiquitously expressed in mammals. After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs (e.g., a2-adrenergic receptor) are phosphorylated by GRKs. Internalization of the GPCRs, regulated by beta-arrestin-1, leads to activation of the Ras - Raf - ERK1&2 signaling pathway. GRK2 activity is tightly controlled by different mechanisms including phosphorylation by kinases such as PKC, Src and ERK1&2, as well as interaction with various proteins. ERK phosphorylates and thus inactivates GRK2 on serine 670 in a negative feedback mechanism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; Beta-ARK-1; EC 2.7.1.126; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2
Gene Aliases: Adrbk-1; ADRBK1; BARK; Bark-1; BARK1; beta ARK; BETA-ARK1; betaARK1; GRK-2; GRK2
UniProt ID: (Mouse) Q99LL8, (Rat) P26817, (Human) P25098
Entrez Gene ID: (Mouse) 110355, (Rat) 25238, (Human) 156
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support