Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESGCFKDINK SESEEALPLD LDKNIHTPVS RPNRGFFYRL FRRGGCLTMV PSEKEVEPKQ
Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 50%.
GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK4 phosphorylates the activated forms of G protein-coupled receptors. GRK4 has been linked to both genetic and acquired hypertension.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor kinase 4; G protein-coupled receptor kinase GRK4; ITI1
Gene Aliases: GPRK2L; GPRK4; GRK4; GRK4a; IT11
UniProt ID: (Human) Q15315
Entrez Gene ID: (Human) 2868
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support