Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Bat, Bovine, Panda, Horse, Pig, Opossum, Turkey, Chicken, Platypus (100%); Orangutan, Dog, Elephant, Xenopus (93%).
For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
GRK5 is a serine/threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-CP4; FP2025; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; GRK; GRK-pan; Pan-GRK; Poly(rC)-binding protein 4
Gene Aliases: 1200003L19Rik; AlphaCP-4; GPRK5; GRK5; Pcbp4
UniProt ID: (Bovine) P43249, (Human) P34947, (Mouse) Q8VEB1, (Rat) Q62833, (Mouse) P57724
Entrez Gene ID: (Bovine) 281801, (Human) 2869, (Chicken) 423929, (Horse) 100065919, (Pig) 100152937, (Mouse) 14773, (Rat) 59075, (Mouse) 59092
Molecular Function:
RNA metabolism protein
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support