Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is K562 cell lysate.
PA5-20547 can be used with blocking peptide PEP-0667.
GRTP1, also known as germ cell-specific gene 1-related protein, is a protein that is expressed in the testis and is involved in the regulation of spermatogenesis, the process of sperm cell development. It is a member of the GRP family of proteins, which are characterized by a conserved motif called the GRP domain. GRTP1 is thought to play a role in the differentiation of spermatogonia into spermatocytes and in the regulation of meiosis, the process of cell division that produces haploid cells. Mutations in the GRTP1 gene have been associated with male infertility. GRTP1 was identified as an HIV dependency factor (HDF), suggesting that GRTP1 may be an important drug target in HIV treatment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GH regulated TBC protein 1; Growth hormone-regulated TBC protein 1; RP11-391H12.7; TBC1 domain family member 6
Gene Aliases: 5430401C05Rik; C81211; GRTP1; LRRGT00052; TBC1D6
UniProt ID: (Human) Q5TC63, (Rat) Q4QQU7, (Mouse) Q9D3N8
Entrez Gene ID: (Human) 79774, (Rat) 361180, (Mouse) 66790
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support