Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAGTSSYW EDLRKQARQL ENELDLKLVS FSKLCTSYSH SSTRDGRRDR YSSDTTPLLN GSSQDRMFET MAIEIEQLLA RLTGVNDKMA EYTNSAGVPS LNAALMHTLQ RHRDILQV (1-117 aa encoded by BC012620)
This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; cis-Golgi p28 (p28); Cis-Golgi SNARE; golgi integral membrane protein 2; Golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOS-28; RP23-343C18.3
Gene Aliases: AI414660; AI426320; BB145494; GOLIM2; GOS-28; GOS28; GOS28/P28; GOSR1; GOSRI; GS28; P28
UniProt ID: (Human) O95249, (Mouse) O88630, (Rat) Q62931
Entrez Gene ID: (Human) 9527, (Mouse) 53334, (Rat) 94189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support