Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%
This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; glutathione reductase; glutathione reductase 1; Glutathione reductase, mitochondrial; glutathione S-reductase; GR; GRase
Gene Aliases: AI325518; D8Ertd238e; GLUR; Gr-1; Gr1; GRD1; GSR; HEL-75; HEL-S-122m; zgc:110010
UniProt ID: (Human) P00390, (Mouse) P47791
Entrez Gene ID: (Human) 2936, (Mouse) 14782, (Zebrafish) 553575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support