Product References
C-terminal conformational changes in SCF-D3/MAX2 ubiquitin ligase are required for KAI2-mediated signaling.
The New phytologist
Tal L,Guercio AM,Varshney K,Young A,Gutjahr C,Shabek N
CAB4169 was used in Western Blot to investigate the function of D3-CTH in the KAR/KL pathway in plants.
Fri Sep 01 00:00:00 UTC 2023
A conformational switch in the SCF-D3/MAX2 ubiquitin ligase facilitates strigolactone signalling.
Nature plants
Tal L,Palayam M,Ron M,Young A,Britt A,Shabek N
CAB4169 was used in Western Blotting to elucidate the functional dynamics of ASK1-D3/MAX2 in SL signalling by leveraging conformational switch mutants in vitro and in plants.
Sun May 01 00:00:00 UTC 2022
Antibodies against human endogenous retrovirus K102 envelope activate neutrophils in systemic lupus erythematosus.
The Journal of experimental medicine
Tokuyama M,Gunn BM,Venkataraman A,Kong Y,Kang I,Rakib T,Townsend MJ,Costenbader KH,Alter G,Iwasaki A
CAB4169 was used in Western Blotting to report the expression of an endogenous retrovirus (ERV) locus ERV-K102, encoding an envelope protein, was significantly elevated in SLE patient blood and correlated with autoantibody levels and higher interferon status.
Mon Jul 05 00:00:00 UTC 2021
Antibodies against human endogenous retrovirus K102 envelope activate neutrophils in systemic lupus erythematosus.
The Journal of experimental medicine
Tokuyama M,Gunn BM,Venkataraman A,Kong Y,Kang I,Rakib T,Townsend MJ,Costenbader KH,Alter G,Iwasaki A
CAB4169 was used in Western Blotting to report the expression of an endogenous retrovirus (ERV) locus ERV-K102, encoding an envelope protein, was significantly elevated in SLE patient blood and correlated with autoantibody levels and higher interferon status.
Mon Jul 05 00:00:00 UTC 2021
Discovery of a Novel CIP2A Variant (NOCIVA) with Clinical Relevance in Predicting TKI Resistance in Myeloid Leukemias.
Clinical cancer research : an official journal of the American Association for Cancer Research
Mäkelä E,Pavic K,Varila T,Salmenniemi U,Löyttyniemi E,Nagelli SG,Ammunét T,Kähäri VM,Clark RE,Elo LL,Bachanaboyina VK,Lucas CM,Itälä-Remes M,Westermarck J
CAB4169 was used in GST Pull Down to discover a novel variant of the oncoprotein CIP2A and its clinical relevance in predicting tyrosine kinase inhibitor therapy resistance in myeloid leukemias.
Sat May 15 00:00:00 UTC 2021
Discovery of a Novel CIP2A Variant (NOCIVA) with Clinical Relevance in Predicting TKI Resistance in Myeloid Leukemias.
Clinical cancer research : an official journal of the American Association for Cancer Research
Mäkelä E,Pavic K,Varila T,Salmenniemi U,Löyttyniemi E,Nagelli SG,Ammunét T,Kähäri VM,Clark RE,Elo LL,Bachanaboyina VK,Lucas CM,Itälä-Remes M,Westermarck J
CAB4169 was used in GST Pull Down to discover a novel variant of the oncoprotein CIP2A and its clinical relevance in predicting tyrosine kinase inhibitor therapy resistance in myeloid leukemias.
Sat May 15 00:00:00 UTC 2021
EBS is a bivalent histone reader that regulates floral phase transition in Arabidopsis.
Nature genetics
Yang Z,Qian S,Scheid RN,Lu L,Chen X,Liu R,Du X,Lv X,Boersma MD,Scalf M,Smith LM,Denu JM,Du J,Zhong X
Published figure using GST Tag polyclonal antibody (Product # CAB4169) in Immunoprecipitation
Sat Sep 01 00:00:00 UTC 2018
EBS is a bivalent histone reader that regulates floral phase transition in Arabidopsis.
Nature genetics
Yang Z,Qian S,Scheid RN,Lu L,Chen X,Liu R,Du X,Lv X,Boersma MD,Scalf M,Smith LM,Denu JM,Du J,Zhong X
Published figure using GST Tag polyclonal antibody (Product # CAB4169) in Immunoprecipitation
Sat Sep 01 00:00:00 UTC 2018
EBS is a bivalent histone reader that regulates floral phase transition in Arabidopsis.
Nature genetics
Yang Z,Qian S,Scheid RN,Lu L,Chen X,Liu R,Du X,Lv X,Boersma MD,Scalf M,Smith LM,Denu JM,Du J,Zhong X
Published figure using GST Tag polyclonal antibody (Product # CAB4169) in Immunoprecipitation
Sat Sep 01 00:00:00 UTC 2018
Dual recognition of H3K4me3 and H3K27me3 by a plant histone reader SHL.
Nature communications
Qian S,Lv X,Scheid RN,Lu L,Yang Z,Chen W,Liu R,Boersma MD,Denu JM,Zhong X,Du J
CAB4169 was used in a Peptide array to establish bromo-adjacent homology-plant homeodomain cassette as a dual histone methyl-lysine binding module that recognizes both active and repressive histone marks.
Thu Jun 21 00:00:00 UTC 2018
Long disordered regions of the C-terminal domain of Abelson tyrosine kinase have specific and additive functions in regulation and axon localization.
PloS one
Cheong HSJ,VanBerkum MFA
CAB4169 was used in Western Blotting to examine the functional role of the long c-terminal domain of Abl.
Thu Jan 04 00:00:00 UTC 2018
Repo-Man/PP1 regulates heterochromatin formation in interphase.
Nature communications
de Castro IJ,Budzak J,Di Giacinto ML,Ligammari L,Gokhan E,Spanos C,Moralli D,Richardson C,de Las Heras JI,Salatino S,Schirmer EC,Ullman KS,Bickmore WA,Green C,Rappsilber J,Lamble S,Goldberg MW,Vinciotti V,Vagnarelli P
CAB4169 was used in western blot to study the role of Repo-Man in interphase
Mon Jan 16 00:00:00 UTC 2017
Breast cancer metastasis suppressor 1 modulates SIRT1-dependent p53 deacetylation through interacting with DBC1.
American journal of cancer research
Liu X,Ehmed E,Li B,Dou J,Qiao X,Jiang W,Yang X,Qiao S,Wu Y
Published figure using GST Tag polyclonal antibody (Product # CAB4169) in Western Blot
Tue Jul 19 00:00:00 UTC 2016
The yeast Rab GTPase Ypt1 modulates unfolded protein response dynamics by regulating the stability of HAC1 RNA.
PLoS genetics
Tsvetanova NG,Riordan DP,Brown PO
Published figure using GST Tag polyclonal antibody (Product # CAB4169) in Western Blot
Fri Jan 04 00:00:00 UTC 2013
Cool-1/βPIX functions as a guanine nucleotide exchange factor in the cycling of Cdc42 to regulate insulin secretion.
American journal of physiology. Endocrinology and metabolism
Kepner EM,Yoder SM,Oh E,Kalwat MA,Wang Z,Quilliam LA,Thurmond DC
CAB4169 was used in immunoprecipitation to identify the interacting partners of Cdc42 and their combined effects on insulin release
Thu Dec 01 00:00:00 UTC 2011
Stimulus-induced S-nitrosylation of Syntaxin 4 impacts insulin granule exocytosis.
The Journal of biological chemistry
Wiseman DA,Kalwat MA,Thurmond DC
CAB4169 was used in western blot to investigate the effect of syntaxin 4 S-nitrosylation on insulin granule exocytosis
Fri May 06 00:00:00 UTC 2011