Product References
Extracellular HSP90 promotes differentiation of lens epithelial cells to fiber cells by activating LRP1-YAP-PROX1 axis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Yu J,Huang W,Sang F,Li J,Ren Y,Huang H,Wang M,Li K,Zhang J,Li H,Cui X,Zhang J,Hu M,Yuan F,Guo W,Zhang F,Mu H,Hu Y
Wed Feb 01 00:00:00 UTC 2023
Extracellular HSP90 promotes differentiation of lens epithelial cells to fiber cells by activating LRP1-YAP-PROX1 axis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Yu J,Huang W,Sang F,Li J,Ren Y,Huang H,Wang M,Li K,Zhang J,Li H,Cui X,Zhang J,Hu M,Yuan F,Guo W,Zhang F,Mu H,Hu Y
Wed Feb 01 00:00:00 UTC 2023
Construction of a Lactobacillus plantarum-based claudin-3 targeting delivery system for the development of vaccines against Eimeria tenella.
Vaccine
Sun L,Zhao N,Li H,Wang B,Li H,Zhang X,Zhao X
Mon Jan 16 00:00:00 UTC 2023
PPARγ inhibited tumor immune escape by inducing PD-L1 autophagic degradation.
Cancer science
Gou Q,Che S,Chen M,Chen H,Shi J,Hou Y
Sat Jul 01 00:00:00 UTC 2023
ERK2 Mutations Affect Interactions, Localization, and Dimerization.
Biochemistry
Taylor CA,Cormier KW,Martin-Vega A,Earnest S,Stippec S,Wichaidit C,Cobb MH
Tue May 02 00:00:00 UTC 2023
FRMD3 inhibits the growth and metastasis of breast cancer through the ubiquitination-mediated degradation of vimentin and subsequent impairment of focal adhesion.
Cell death & disease
Shao W,Li J,Piao Q,Yao X,Li M,Wang S,Song Z,Sun Y,Zheng L,Wang G,Liu L,Yu C,Huang Y,Bao Y,Sun L
Wed Jan 11 00:00:00 UTC 2023
FRMD3 inhibits the growth and metastasis of breast cancer through the ubiquitination-mediated degradation of vimentin and subsequent impairment of focal adhesion.
Cell death & disease
Shao W,Li J,Piao Q,Yao X,Li M,Wang S,Song Z,Sun Y,Zheng L,Wang G,Liu L,Yu C,Huang Y,Bao Y,Sun L
Wed Jan 11 00:00:00 UTC 2023
SEPT2 crotonylation promotes metastasis and recurrence in hepatocellular carcinoma and is associated with poor survival.
Cell & bioscience
Zhang XY,Liu ZX,Zhang YF,Xu LX,Chen MK,Zhou YF,Yu J,Li XX,Zhang N
Wed Mar 22 00:00:00 UTC 2023
Silencing a Chitinase Gene, PstChia1, Reduces Virulence of Puccinia striiformis f. sp. tritici.
International journal of molecular sciences
Guo J,Mou Y,Li Y,Yang Q,Wang X,Lin H,Kang Z,Guo J
Thu May 04 00:00:00 UTC 2023
N-homocysteinylation of α-synuclein promotes its aggregation and neurotoxicity.
Aging cell
Zhou L,Guo T,Meng L,Zhang X,Tian Y,Dai L,Niu X,Li Y,Liu C,Chen G,Liu C,Ke W,Zhang Z,Bao A,Zhang Z
Wed Mar 01 00:00:00 UTC 2023
Stabilization of estrogen receptor α by USP37 contributes to the progression of breast cancer.
Cancer science
Cao J,Wang X,Wang S,Chen Z,Tang J
Mon May 01 00:00:00 UTC 2023
Bre1/RNF20 promotes Rad51-mediated strand exchange and antagonizes the Srs2/FBH1 helicases.
Nature communications
Liu G,Li J,He B,Yan J,Zhao J,Wang X,Zhao X,Xu J,Wu Y,Zhang S,Gan X,Zhou C,Li X,Zhang X,Chen X
Thu May 25 00:00:00 UTC 2023
Bre1/RNF20 promotes Rad51-mediated strand exchange and antagonizes the Srs2/FBH1 helicases.
Nature communications
Liu G,Li J,He B,Yan J,Zhao J,Wang X,Zhao X,Xu J,Wu Y,Zhang S,Gan X,Zhou C,Li X,Zhang X,Chen X
Thu May 25 00:00:00 UTC 2023
Bacterial infection promotes tumorigenesis of colorectal cancer via regulating CDC42 acetylation.
PLoS pathogens
Wang DN,Ni JJ,Li JH,Gao YQ,Ni FJ,Zhang ZZ,Fang JY,Lu J,Yao YF
Wed Feb 01 00:00:00 UTC 2023
Sensing of a spore surface protein by a Drosophila chemosensory protein induces behavioral defense against fungal parasitic infections.
Current biology : CB
Shang J,Tang G,Yang J,Lu M,Wang CZ,Wang C
Mon Jan 23 00:00:00 UTC 2023
The Valsa Mali effector Vm1G-1794 protects the aggregated MdEF-Tu from autophagic degradation to promote infection in apple.
Autophagy
Che R,Liu C,Wang Q,Tu W,Wang P,Li C,Gong X,Mao K,Feng H,Huang L,Li P,Ma F
Thu Jun 01 00:00:00 UTC 2023
FOXM1 increases hTERT protein stability and indicates poor prognosis in gastric cancer.
Neoplasia (New York, N.Y.)
Tang Q,Liu C,Zhang S,He L,Liu Y,Wang J,Zhao X,Li X
Wed Feb 01 00:00:00 UTC 2023
The moonlighting function of glycolytic enzyme enolase-1 promotes choline phospholipid metabolism and tumor cell proliferation.
Proceedings of the National Academy of Sciences of the United States of America
Ma Q,Jiang H,Ma L,Zhao G,Xu Q,Guo D,He N,Liu H,Meng Z,Liu J,Zhu L,Lin Q,Wu X,Li M,Luo S,Fang J,Lu Z
Tue Apr 11 00:00:00 UTC 2023
Cofilin promotes tau pathology in Alzheimer's disease.
Cell reports
Yan M,Tang L,Dai L,Lei C,Xiong M,Zhang X,He M,Tian Y,Xiong J,Ke W,Zhang Z,Zhang C,Deng X,Zhang Z
Tue Feb 28 00:00:00 UTC 2023
OsNF-YA3 regulates plant growth and osmotic stress tolerance by interacting with SLR1 and SAPK9 in rice.
The Plant journal : for cell and molecular biology
Jin X,Zhang Y,Li X,Huang J
Mon May 01 00:00:00 UTC 2023
Elevated Nuclear PHGDH Synergistically Functions with cMyc to Reshape the Immune Microenvironment of Liver Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhu H,Yu H,Zhou H,Zhu W,Wang X
Thu Jun 01 00:00:00 UTC 2023
Elevated Nuclear PHGDH Synergistically Functions with cMyc to Reshape the Immune Microenvironment of Liver Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhu H,Yu H,Zhou H,Zhu W,Wang X
Thu Jun 01 00:00:00 UTC 2023
Islet amyloid polypeptide triggers α-synuclein pathology in Parkinson's disease.
Progress in neurobiology
Meng L,Li Y,Liu C,Zhang G,Chen J,Xiong M,Pan L,Zhang X,Chen G,Xiong J,Liu C,Xu X,Bu L,Zhang Z,Zhang Z
Sat Jul 01 00:00:00 UTC 2023
The deubiquitinase OTUD1 noncanonically suppresses Akt activation through its N-terminal intrinsically disordered region.
Cell reports
Fan G,Wang F,Chen Y,Zheng Q,Xiong J,Lv Q,Wu K,Xiong J,Wei L,Li D,Zhang J,Zhang W,Li F
Tue Jan 31 00:00:00 UTC 2023
The deubiquitinase OTUD1 noncanonically suppresses Akt activation through its N-terminal intrinsically disordered region.
Cell reports
Fan G,Wang F,Chen Y,Zheng Q,Xiong J,Lv Q,Wu K,Xiong J,Wei L,Li D,Zhang J,Zhang W,Li F
Tue Jan 31 00:00:00 UTC 2023
7,8-Dihydroxyflavone ameliorates mitochondrial impairment and motor dysfunction in the α-synuclein 1-103 transgenic mice.
Neurobiology of disease
Tian Y,Pan L,Yuan X,Xiong M,Zhang Z,Meng L,Zheng Y,Bu L,Xu X,Zhang Z
Fri Jul 01 00:00:00 UTC 2022
Aerobic glycolysis promotes tumor immune evasion by hexokinase2-mediated phosphorylation of IκBα.
Cell metabolism
Guo D,Tong Y,Jiang X,Meng Y,Jiang H,Du L,Wu Q,Li S,Luo S,Li M,Xiao L,He H,He X,Yu Q,Fang J,Lu Z
Tue Sep 06 00:00:00 UTC 2022
Insights into acylation mechanisms: co-expression of serine carboxypeptidase-like acyltransferases and their non-catalytic companion paralogs.
The Plant journal : for cell and molecular biology
Yao S,Liu Y,Zhuang J,Zhao Y,Dai X,Jiang C,Wang Z,Jiang X,Zhang S,Qian Y,Tai Y,Wang Y,Wang H,Xie DY,Gao L,Xia T
Fri Jul 01 00:00:00 UTC 2022
The metalloprotease ADM-4/ADAM17 promotes axonal repair.
Science advances
Ho XY,Coakley S,Amor R,Anggono V,Hilliard MA
Fri Mar 18 00:00:00 UTC 2022
G Protein Subunit β1 Facilitates Influenza A Virus Replication by Promoting the Nuclear Import of PB2.
Journal of virology
Zheng H,Ma L,Gui R,Lin X,Ke X,Jian X,Ye C,Chen Q
Wed Jun 22 00:00:00 UTC 2022
Rice OsPUB16 modulates the 'SAPK9-OsMADS23-OsAOC' pathway to reduce plant water-deficit tolerance by repressing ABA and JA biosynthesis.
PLoS genetics
Lv Q,Li X,Jin X,Sun Y,Wu Y,Wang W,Huang J
Tue Nov 01 00:00:00 UTC 2022
Rice OsPUB16 modulates the 'SAPK9-OsMADS23-OsAOC' pathway to reduce plant water-deficit tolerance by repressing ABA and JA biosynthesis.
PLoS genetics
Lv Q,Li X,Jin X,Sun Y,Wu Y,Wang W,Huang J
Tue Nov 01 00:00:00 UTC 2022
Legionella pneumophila regulates host cell motility by targeting Phldb2 with a 14-3-3ζ-dependent protease effector.
eLife
Song L,Luo J,Wang H,Huang D,Tan Y,Liu Y,Wang Y,Yu K,Zhang Y,Liu X,Li D,Luo ZQ
Thu Feb 17 00:00:00 UTC 2022
Auxin and abscisic acid antagonistically regulate ascorbic acid production via the SlMAPK8-SlARF4-SlMYB11 module in tomato.
The Plant cell
Xu X,Zhang Q,Gao X,Wu G,Wu M,Yuan Y,Zheng X,Gong Z,Hu X,Gong M,Qi T,Li H,Luo Z,Li Z,Deng W
Thu Oct 27 00:00:00 UTC 2022
Elevated FAM84B promotes cell proliferation via interacting with NPM1 in esophageal squamous cell carcinoma.
Cell death discovery
Wang F,Cheng C,Wang X,Chen F,Li H,Zhou Y,Wang Y,Hu X,Kong P,Zhang L,Cheng X,Cui Y
Fri Apr 08 00:00:00 UTC 2022
Skin chronological aging drives age-related bone loss via secretion of cystatin-A.
Nature aging
Liang W,Chen Q,Cheng S,Wei R,Li Y,Yao C,Ouyang Z,Kang D,Chen A,Liu Z,Li K,Bai X,Li Q,Huang B
Sat Oct 01 00:00:00 UTC 2022
circPRKAA1 activates a Ku80/Ku70/SREBP-1 axis driving de novo fatty acid synthesis in cancer cells.
Cell reports
Li Q,Yao H,Wang Y,Wu Y,Thorne RF,Zhu Y,Wu M,Liu L
Tue Nov 22 00:00:00 UTC 2022
Aurora A-mediated pyruvate kinase M2 phosphorylation promotes biosynthesis with glycolytic metabolites and tumor cell cycle progression.
The Journal of biological chemistry
Jiang Y,Wang T,Sheng D,Han C,Xu T,Zhang P,You W,Fan W,Zhang Z,Jin T,Duan X,Yuan X,Liu X,Zhang K,Ruan K,Shi J,Guo J,Cheng A,Yang Z
Tue Nov 01 00:00:00 UTC 2022
The histone methyltransferase SUVR2 promotes DSB repair via chromatin remodeling and liquid-liquid phase separation.
Molecular plant
Liu Q,Liu P,Ji T,Zheng L,Shen C,Ran S,Liu J,Zhao Y,Niu Y,Wang T,Dong J
Mon Jul 04 00:00:00 UTC 2022
The histone methyltransferase SUVR2 promotes DSB repair via chromatin remodeling and liquid-liquid phase separation.
Molecular plant
Liu Q,Liu P,Ji T,Zheng L,Shen C,Ran S,Liu J,Zhao Y,Niu Y,Wang T,Dong J
Mon Jul 04 00:00:00 UTC 2022
METTL16 antagonizes MRE11-mediated DNA end resection and confers synthetic lethality to PARP inhibition in pancreatic ductal adenocarcinoma.
Nature cancer
Zeng X,Zhao F,Cui G,Zhang Y,Deshpande RA,Chen Y,Deng M,Kloeber JA,Shi Y,Zhou Q,Zhang C,Hou J,Kim W,Tu X,Yan Y,Xu Z,Chen L,Gao H,Guo G,Liu J,Zhu Q,Cao Y,Huang J,Wu Z,Zhu S,Yin P,Luo K,Mer G,Paull TT,Yuan J,Tao K,Lou Z
Thu Sep 01 00:00:00 UTC 2022
A synapsin ¿ cleavage fragment contributes to synaptic dysfunction in Alzheimer's disease.
Aging cell
Meng L,Zou L,Xiong M,Chen J,Zhang X,Yu T,Li Y,Liu C,Chen G,Wang Z,Ye K,Zhang Z
Sun May 01 00:00:00 UTC 2022
A novel protein encoded by ZCRB1-induced circHEATR5B suppresses aerobic glycolysis of GBM through phosphorylation of JMJD5.
Journal of experimental & clinical cancer research : CR
Song J,Zheng J,Liu X,Dong W,Yang C,Wang D,Ruan X,Zhao Y,Liu L,Wang P,Zhang M,Liu Y
Tue May 10 00:00:00 UTC 2022
Ivermectin represses Wnt/β-catenin signaling by binding to TELO2, a regulator of phosphatidylinositol 3-kinase-related kinases.
iScience
Yonezawa H,Ikeda A,Takahashi R,Endo H,Sugawara Y,Goto M,Kanno M,Ogawa S,Nakamura K,Ujiie H,Iwatsuki M,Hirose T,Sunazuka T,Uehara Y,Nishiya N
Fri Mar 18 00:00:00 UTC 2022
Ciliary Type III Adenylyl Cyclase in the VMH Is Crucial for High-Fat Diet-Induced Obesity Mediated by Autophagy.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Yang D,Wu X,Wang W,Zhou Y,Wang Z
Sat Jan 01 00:00:00 UTC 2022
Claudin-5 binder enhances focused ultrasound-mediated opening in an in vitro blood-brain barrier model.
Theranostics
Chen L,Sutharsan R,Lee JL,Cruz E,Asnicar B,Palliyaguru T,Wasielewska JM,Gaudin A,Song J,Leinenga G,Götz J
Thu Apr 21 00:00:00 UTC 2022
Evolutionary Analysis and Functional Identification of Ancient Brassinosteroid Receptors in Ceratopteris richardii.
International journal of molecular sciences
Zheng B,Xing K,Zhang J,Liu H,Ali K,Li W,Bai Q,Ren H
Sat Jun 18 00:00:00 UTC 2022
Transcription factors ZmNF-YA1 and ZmNF-YB16 regulate plant growth and drought tolerance in maize.
Plant physiology
Yang Y,Wang B,Wang J,He C,Zhang D,Li P,Zhang J,Li Z
Wed Sep 28 00:00:00 UTC 2022
The tomato OST1-VOZ1 module regulates drought-mediated flowering.
The Plant cell
Chong L,Xu R,Huang P,Guo P,Zhu M,Du H,Sun X,Ku L,Zhu JK,Zhu Y
Tue Apr 26 00:00:00 UTC 2022
The histone deacetylase 1/GSK3/SHAGGY-like kinase 2/BRASSINAZOLE-RESISTANT 1 module controls lateral root formation in rice.
Plant physiology
Hou J,Zheng X,Ren R,Shi Q,Xiao H,Chen Z,Yue M,Wu Y,Hou H,Li L
Wed Jun 01 00:00:00 UTC 2022
Islet amyloid polypeptide cross-seeds tau and drives the neurofibrillary pathology in Alzheimer's disease.
Molecular neurodegeneration
Zhang G,Meng L,Wang Z,Peng Q,Chen G,Xiong J,Zhang Z
Sat Jan 29 00:00:00 UTC 2022
Linear ubiquitination of PTEN impairs its function to promote prostate cancer progression.
Oncogene
Guo Y,He J,Zhang H,Chen R,Li L,Liu X,Huang C,Qiang Z,Zhou Z,Wang Y,Huang J,Zhao X,Zheng J,Chen GQ,Yu J
Sat Oct 01 00:00:00 UTC 2022
The Therapeutic Roles of Recombinant Hsp90α on Cornea Epithelial Injury.
Investigative ophthalmology & visual science
Wang M,Hu J,Qu J,Huang H,Zhang J,Zhang J,Li H,Cui X,Zhang F,Hu MY,Li J,Hu Y
Tue Feb 01 00:00:00 UTC 2022
The Therapeutic Roles of Recombinant Hsp90α on Cornea Epithelial Injury.
Investigative ophthalmology & visual science
Wang M,Hu J,Qu J,Huang H,Zhang J,Zhang J,Li H,Cui X,Zhang F,Hu MY,Li J,Hu Y
Tue Feb 01 00:00:00 UTC 2022
OsNAC016 regulates plant architecture and drought tolerance by interacting with the kinases GSK2 and SAPK8.
Plant physiology
Wu Q,Liu Y,Xie Z,Yu B,Sun Y,Huang J
Mon Jun 27 00:00:00 UTC 2022
A γ-adducin cleavage fragment induces neurite deficits and synaptic dysfunction in Alzheimer's disease.
Progress in neurobiology
Xiong M,Zou L,Meng L,Zhang X,Tian Y,Zhang G,Yang J,Chen G,Xiong J,Ye K,Zhang Z
Sun Aug 01 00:00:00 UTC 2021
Amphiphysin I cleavage by asparagine endopeptidase leads to tau hyperphosphorylation and synaptic dysfunction.
eLife
Zhang X,Zou L,Meng L,Xiong M,Pan L,Chen G,Zheng Y,Xiong J,Wang Z,Duong DM,Zhang Z,Cao X,Wang T,Tang L,Ye K,Zhang Z
Fri May 21 00:00:00 UTC 2021
Regulation of NMDA receptor trafficking and gating by activity-dependent CaMKIIα phosphorylation of the GluN2A subunit.
Cell reports
Yong XLH,Zhang L,Yang L,Chen X,Tan JZA,Yu X,Chandra M,Livingstone E,Widagdo J,Vieira MM,Roche KW,Lynch JW,Keramidas A,Collins BM,Anggono V
Tue Jul 06 00:00:00 UTC 2021
Identification of novel B-cell epitopes on the capsid protein of type 1 porcine astrovirus, using monoclonal antibodies.
International journal of biological macromolecules
Zhang W,Wang W,Liu X,Chen Y,Ouyang K,Wei Z,Liu H,Huang W
Sun Oct 31 00:00:00 UTC 2021
Autophagy Induced by the N-Terminus of the Classic Swine Fever Virus Nonstructural Protein 5A Protein Promotes Viral Replication.
Frontiers in microbiology
Zhang C,Wang X,Sun J,Guo M,Zhang X,Wu Y
Tue Sep 14 00:00:00 UTC 2021
Autophagy Induced by the N-Terminus of the Classic Swine Fever Virus Nonstructural Protein 5A Protein Promotes Viral Replication.
Frontiers in microbiology
Zhang C,Wang X,Sun J,Guo M,Zhang X,Wu Y
Tue Sep 14 00:00:00 UTC 2021
Kinase Function of Brassinosteroid Receptor Specified by Two Allosterically Regulated Subdomains.
Frontiers in plant science
Ali K,Li W,Qin Y,Wang S,Feng L,Wei Q,Bai Q,Zheng B,Li G,Ren H,Wu G
Tue Feb 01 00:00:00 UTC 2022
Identification of the Bcl-2 and Bax homologs from Rhipicephalus haemaphysaloides and their function in the degeneration of tick salivary glands.
Parasites & vectors
Hu S,Wang Y,Xu Z,Zhou Y,Cao J,Zhang H,Zhou J
Wed Aug 04 00:00:00 UTC 2021
Maize transcription factor ZmNF-YC13 regulates plant architecture.
Journal of experimental botany
Mei X,Nan J,Zhao Z,Yao S,Wang W,Yang Y,Bai Y,Dong E,Liu C,Cai Y
Tue Jun 22 00:00:00 UTC 2021
Asparagine endopeptidase cleaves synaptojanin 1 and triggers synaptic dysfunction in Parkinson's disease.
Neurobiology of disease
Zou L,Zhang X,Xiong M,Meng L,Tian Y,Pan L,Yuan X,Chen G,Wang Z,Bu L,Yao Z,Zhang Z,Ye K,Zhang Z
Thu Jul 01 00:00:00 UTC 2021
LRRC4 mediates the formation of circular RNA CD44 to inhibitGBM cell proliferation.
Molecular therapy. Nucleic acids
Feng J,Ren X,Fu H,Li D,Chen X,Zu X,Liu Q,Wu M
Fri Dec 03 00:00:00 UTC 2021
LRRC4 mediates the formation of circular RNA CD44 to inhibitGBM cell proliferation.
Molecular therapy. Nucleic acids
Feng J,Ren X,Fu H,Li D,Chen X,Zu X,Liu Q,Wu M
Fri Dec 03 00:00:00 UTC 2021
Inhibition of AXL enhances chemosensitivity of human ovarian cancer cells to cisplatin via decreasing glycolysis.
Acta pharmacologica Sinica
Tian M,Chen XS,Li LY,Wu HZ,Zeng D,Wang XL,Zhang Y,Xiao SS,Cheng Y
Thu Jul 01 00:00:00 UTC 2021
Inhibition of AXL enhances chemosensitivity of human ovarian cancer cells to cisplatin via decreasing glycolysis.
Acta pharmacologica Sinica
Tian M,Chen XS,Li LY,Wu HZ,Zeng D,Wang XL,Zhang Y,Xiao SS,Cheng Y
Thu Jul 01 00:00:00 UTC 2021
STAT3-Dependent Gene TRIM5γ Interacts With HBx Through a Zinc Binding Site on the BBox Domain.
Frontiers in microbiology
Song H,Xu F,Pang X,Xiao Q,Wei Q,Lei B,Li X,Fan X,Tan G
Tue Jul 20 00:00:00 UTC 2021
Site-specific SUMOylation of viral polymerase processivity factor: a way of localizingtoND10 subnuclear domains for restricted and self-controlled reproduction of herpesvirus.
Virulence
Lai S,Xu M,Wang Y,Li R,Xia C,Xia S,Chen J
Wed Dec 01 00:00:00 UTC 2021
Bach1-induced suppression of angiogenesis is dependent on the BTB domain.
EBioMedicine
Jiang L,Jia M,Wei X,Guo J,Hao S,Mei A,Zhi X,Wang X,Li Q,Jin J,Zhang J,Li S,Meng D
Wed Jan 01 00:00:00 UTC 2020
Dehydrin MtCAS31 promotes autophagic degradation under drought stress.
Autophagy
Li X,Liu Q,Feng H,Deng J,Zhang R,Wen J,Dong J,Wang T
Fri May 01 00:00:00 UTC 2020
Fine mapping of linear B cell epitopes on capsid protein of porcine circovirus 3.
Applied microbiology and biotechnology
Jiang M,Guo J,Zhang G,Jin Q,Liu Y,Jia R,Wang A
Wed Jul 01 00:00:00 UTC 2020
Heat shock factor 4 regulates lysosome activity by modulating the αB-crystallin-ATP6V1A-mTOR complex in ocular lens.
Biochimica et biophysica acta. General subjects
Cui X,Feng R,Wang J,Du C,Pi X,Chen D,Li J,Li H,Zhang J,Zhang J,Mu H,Zhang F,Liu M,Hu Y
Sun Mar 01 00:00:00 UTC 2020
Proto-oncogene Src links lipogenesis via lipin-1 to breast cancer malignancy.
Nature communications
Song L,Liu Z,Hu HH,Yang Y,Li TY,Lin ZZ,Ye J,Chen J,Huang X,Liu DT,Zhou J,Shi Y,Zhao H,Xie C,Chen L,Song E,Lin SY,Lin SC
Tue Nov 17 00:00:00 UTC 2020
PICK1 Controls Activity-Dependent Synaptic Vesicle Cargo Retrieval.
Cell reports
Yong XLH,Cousin MA,Anggono V
Tue Oct 27 00:00:00 UTC 2020
FGF15 Activates Hippo Signaling to Suppress Bile Acid Metabolism and Liver Tumorigenesis.
Developmental cell
Ji S,Liu Q,Zhang S,Chen Q,Wang C,Zhang W,Xiao C,Li Y,Nian C,Li J,Li J,Geng J,Hong L,Xie C,He Y,Chen X,Li X,Yin ZY,You H,Lin KH,Wu Q,Yu C,Johnson RL,Wang L,Chen L,Wang F,Zhou D
Mon Feb 25 00:00:00 UTC 2019
Nonmuscle Myosin Heavy Chain IIA Recognizes Sialic Acids on Sialylated RNA Viruses To Suppress Proinflammatory Responses via the DAP12-Syk Pathway.
mBio
Liu Y,Li R,Chen XX,Zhi Y,Deng R,Zhou EM,Qiao S,Zhang G
Tue May 07 00:00:00 UTC 2019
Phagolysosome resolution requires contacts with the endoplasmic reticulum and phosphatidylinositol-4-phosphate signalling.
Nature cell biology
Levin-Konigsberg R,Montaño-Rendón F,Keren-Kaplan T,Li R,Ego B,Mylvaganam S,DiCiccio JE,Trimble WS,Bassik MC,Bonifacino JS,Fairn GD,Grinstein S
Tue Oct 01 00:00:00 UTC 2019
The galectin LEC-5 is a novel binding partner for RAB-11.
Biochemical and biophysical research communications
Zhang N,Wang X,Gobel V,Zhang X
Sun Oct 28 00:00:00 UTC 2018
The galectin LEC-5 is a novel binding partner for RAB-11.
Biochemical and biophysical research communications
Zhang N,Wang X,Gobel V,Zhang X
Sun Oct 28 00:00:00 UTC 2018
Characterization of a novel secretory spherical body protein in Babesia orientalis and Babesia orientalis-infected erythrocytes.
Parasites & vectors
Guo J,Li M,Sun Y,Yu L,He P,Nie Z,Zhan X,Zhao Y,Luo X,Wang S,Aoyang S,Liu Q,Huang C,He L,Zhao J
Wed Jul 25 00:00:00 UTC 2018
Characterization of a novel secretory spherical body protein in Babesia orientalis and Babesia orientalis-infected erythrocytes.
Parasites & vectors
Guo J,Li M,Sun Y,Yu L,He P,Nie Z,Zhan X,Zhao Y,Luo X,Wang S,Aoyang S,Liu Q,Huang C,He L,Zhao J
Wed Jul 25 00:00:00 UTC 2018
Characterization of a novel secretory spherical body protein in Babesia orientalis and Babesia orientalis-infected erythrocytes.
Parasites & vectors
Guo J,Li M,Sun Y,Yu L,He P,Nie Z,Zhan X,Zhao Y,Luo X,Wang S,Aoyang S,Liu Q,Huang C,He L,Zhao J
Wed Jul 25 00:00:00 UTC 2018
Characterization of a novel secretory spherical body protein in Babesia orientalis and Babesia orientalis-infected erythrocytes.
Parasites & vectors
Guo J,Li M,Sun Y,Yu L,He P,Nie Z,Zhan X,Zhao Y,Luo X,Wang S,Aoyang S,Liu Q,Huang C,He L,Zhao J
Wed Jul 25 00:00:00 UTC 2018