Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene encodes a member of a family of enzymes that function to add glutathione to target electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress. This action is an important step in detoxification of these compounds. This subfamily of enzymes has a particular role in protecting cells from reactive oxygen species and the products of peroxidation. Polymorphisms in this gene influence the ability of individuals to metabolize different drugs. This gene is located in a cluster of similar genes and pseudogenes on chromosome 6. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; class alpha; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase 2; Glutathione S-transferase A1; glutathione S-transferase A3 subunit; glutathione S-transferase A5; Glutathione S-transferase alpha-3; glutathione S-transferase Ha subunit 1; Glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; Glutathione-S-transferase alpha type (Ya); GST 2-2; GST A3-3; GST AA; GST class-alpha member 1; GST HA subunit 1; GST Yc1; GST-epsilon; GSTA1-1; GTH1; RP11-392A23.1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80
Gene Aliases: GST-epsilon; GST2; GSTA1; GSTA1-1; Gsta3; Gsta5; Gstyc1; GTH1; Yc1
UniProt ID: (Human) P08263, (Rat) P04904
Entrez Gene ID: (Human) 2938, (Rat) 24421
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support