Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGPLPRTVE LFYDVLSPYS WLGFEILCRY QNIWNINLQL RPSLITGIMK DSGNKPPGLL PRKGLYMAND LKLLRHHLQI PIHFPKDFLS VMLEKGSLSA MRFLTAVNLE HPEMLEKASR ELWMRVWSRN EDITEPQSIL AAAEKAGMSA EQAQGLLEKI ATPKVKNQLK ETTEAACRYG AFGLPITVAH VDGQTHMLFG SDRMELLAHL LGEKWMGPIP PAVNARL (1-226 aa encoded by BC050715)
GSTK1 encodes a member of the kappa class of the glutathione transferase superfamily of enzymes that function in cellular detoxification. The encoded protein is localized to the peroxisome and catalyzes the conjugation of glutathione to a wide range of hydrophobic substates facilitating the removal of these compounds from cells. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutathione S-transferase class kappa; glutathione S-transferase k1; Glutathione S-transferase kappa 1; Glutathione S-transferase subunit 13; glutathione S-transferase subunit 13 homolog; glutathione S-transferase, mitochondrial; GST 13-13; GST class-kappa; GSTK1-1; hGSTK1; mGSTK1; rGSTK1
Gene Aliases: 0610025I19Rik; AW260476; DsbA-L; GST; GST 13-13; GST13; GST13-13; GSTK1; GSTK1-1; GSTkappa; HDCMD47P; hGSTK1
UniProt ID: (Human) Q9Y2Q3, (Mouse) Q9DCM2, (Rat) P24473
Entrez Gene ID: (Human) 373156, (Mouse) 76263, (Rat) 297029
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support