Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSILIIEAFY GGSHKQLVDL LQEELGDCVV YTLPAKKWHW RARTSALYFS QTIPISEHYR TLFASSVLNL TELAALRPDL GKLKKILYFH ENQLIYPVKK CQERDFQYGY NQILSCLVAD VVVFNSVFNM ESFLTSMGKF MKLIPDHRPK DLESIIRPKC QVIYFPIRFP DVSRFMPKHK TTHLKKMLGL KGNGGAVLSM ALPFQPEQRD SEDLLKNFNS ECDTHCGLDT ARQEYLGNSL RQESDLKKST SSDNSSSHHG ENKQNLTVDP CDILGGVDNQ QRLLHIVWPH RWLEAVYCGC YPLCPKDLVY PEIFPAEYLY STPEQLSKRL QNFCKRPDII RKHLYKGEIA PFSWAALHGK FRSLLTTEPR EDL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycosyltransferase-like 1; glycosyltransferase-like domain containing 1; Glycosyltransferase-like domain-containing protein 1; mannosyltransferase candidate; Mannosyltransferase-like protein Xa; Mat-Xa; QTMAN; tRNA-queuosine alpha-mannosyltransferase
Gene Aliases: GTDC1; Hmat-Xa; mat-Xa; QTMAN
UniProt ID: (Human) Q4AE62
Entrez Gene ID: (Human) 79712
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support