Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QIIHFLRTRA HPVMLKQTPV LPPTITDQIR LWELERDRLR FTEGVLYNQF LSQVDFELLL AHARELGVLV FENSAKRLMV VTPAGHSDVK RFWKRQKHSS
GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic transcription factor 2 52 kDa subunit; BTF2 p52; General transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; TFIIH basal transcription factor complex p52 subunit
Gene Aliases: GTF2H4; P52; TFB2; TFIIH
UniProt ID: (Human) Q92759
Entrez Gene ID: (Human) 2968
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support