Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
It is not recommended to aliquot this antibody.
This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAP-135; Bruton tyrosine kinase-associated protein 135; BTK-associated protein 135; BTK-associated protein, 135kD; General transcription factor II-I; GTFII-I; OTTHUMP00000209509; SPIN; SRF-Phox1-interacting protein; tfiii; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6
Gene Aliases: 6030441I21Rik; BAP-135; BAP135; BTKAP1; DIWS; Diws1t; GTF2I; GTFII-I; IB291; SPIN; TFII-I; WBS; WBSCR6
UniProt ID: (Human) P78347, (Mouse) Q9ESZ8
Entrez Gene ID: (Human) 2969, (Mouse) 14886
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support