Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALLGKRCDV PTNGCGPDRW NSAFTRKDEI ITSLVSALDS MCSALSKLNA EVACVAVHDE SAFVVGTEKG RMFLNARKEL QSDFLRFCRG PPWKDPEAEH PKKVQRGEGG GRSLPRSSLE HGSDVYLLRK MVEEVFDVLY SEALGRASVV PLPYERLLRE PGLLAVQGLP EGLAFRRPAE YDPKALMAIL EHSHRIRFKL KRPLEDGGRD SKALVELNGV SLIPKGSRDC GLHGQAPKVP PQDLPPTATS SSMASFLYST ALPNHAIREL KQEAPSCPLA PSDLGLSRPM PEPKATGAQD FSDCCGQKPT GPGGPLIQNV HASKRILFSI VHDKSEKWDA FIKETEDINT LRECVQILFN SRYAEALGLD HMVPVPYRKI ACDPEAVEIV GIPDKIPFKR PCTYGVPKLK RILEERHSIH FIIKRMFDER IFTGNKFTKD TTKLEPASPP EDTSAEVSRA TVLDLAGNAR SDKGSMSEDC GPGTSGELGG LRPIKIEPED LDIIQVTVPD PSPTSEEMTD SMPGHLPSED SGYGMEMLTD KGLSEDARPE ERPVEDSHGD VIRPLRKQVE LLFNTRYAKA IGISEPVKVP YSKFLMHPEE LFVVGLPEGI SLRRPNCFGI AKLRKILEAS NSIQFVIKRP ELLTEGVKEP IVDSQGTASS LGFSPPALPP ERDSGDPLVD ESLKRQGFQE NYDARLSRID IANTLREQVQ DLFNKKYGEA LGIKYPVQVP YKRIKSNPGS VIIEGLPPGI PFRKPCTFGS QNLERILAVA DKIKFTVTRP FQGLIPKPDE DDANRLGEKV ILREQVKELF NEKYGEALGL NRPVLVPYKL IRDSPDAVEV TGLPDDIPFR NPNTYDIHRL EKILKAREHV RMVIINQLQP FAEICNDAKV PAKDSSIPKR KRKRVSEGNS VSSSSSSSSS SSSNPDSVAS ANQISLVQWP MYMVDYAGLN VQLPGPLNY
Williams-Beuren syndrome (WBS) is a developmental disorder caused by the hemizygous microdeletion on chromosome 7q11.23. WBS is an autosomal dominant genetic condition that is characterized by physical, cognitive and behavioral traits. The physical traits associated with WBS include facial dysmorphology, vascular stenoses, growth deficiencies, dental anomalies and neurologic and musculoskeletal abnormalities. Mild retardation, a weakness in visual-spatial skills, anxiety and a short attention span are typical cognitive and behavioral traits of WBS patients. The WBSCR11 gene is located within the WBS deletion and may contribute to the developmental symptoms found in WBS because of a loss of the encoded transcription factor. WBSCR11 is also designated GRF2IRD1, GTF3, Cream1 and MusTRD1 in human and BEN in mouse, due to slight differences in gene structure. WBSCR11 is expressed in all adult tissues as several variants and has discrete spatial and temporal expression during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: binding factor for early enhancer; general transcription factor 3; General transcription factor II-I repeat domain-containing protein 1; General transcription factor III; GTF2I repeat domain-containing protein 1; Muscle TFII-I repeat domain-containing protein 1; muscle TFII-I repeat domain-containing protein 1 alpha 1; MusTRD1/BEN; Slow-muscle-fiber enhancer-binding protein; USE B1-binding protein; Williams-Beuren syndrome chromosomal region 11 protein; Williams-Beuren syndrome chromosomal region 12 protein; Williams-Beuren syndrome chromosome region 11
Gene Aliases: BEN; CREAM1; GTF2IRD1; GTF3; hMusTRD1alpha1; MUSTRD1; RBAP2; WBS; WBSCR11; WBSCR12
UniProt ID: (Human) Q9UHL9
Entrez Gene ID: (Human) 9569
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support