Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAEAADLGL GAAVPVELRR ERRMVCVEYP GVVRDVAKML PTLGGEEGVS RIYADPTKRL ELYFRPKDPY CHPVCANRFS TSSLLLRIRK RTRRQKGVLG TEAHSEVTFD MEILGIISTI YKFQGMSDFQ YLAVHTEAGG KHTSMYDKVL MLRPEKEAFF HQELPLYIPP PIFSRLDAPV DYFYRPETQH REGYNNPPIS GENLIGLSRA RRPHNAIFVN FEDEEVPKQP LEAAAQTWRR VCTNPVDRKV EEELRKLFDI RPIWSRNAVK ANISVHPDKL KVLLPFIAYY MITGPWRSLW IRFGYDPRKN PDAKIYQVLD FRIRCGMKHG YAPSDLPVKA KRSTYNYSLP ITVKKTSSQL VTMHDLKQGL GPSGTSGARK PASSKYKLKD SVYIFREGAL PPYRQMFYQL CDLNVEELQK IIHRNDGAEN SCTERDGWCL PKTSDELRDT MSLMIRQTIR SKRPALFSSS AKADGGKEQL TYESGEDEED EEEEEEEEED FKPSDGSENE METEILDYV
Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: General transcription factor 3C polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; TF3C-epsilon; TFIIIC 63 kDa subunit; Transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD
Gene Aliases: CDABP0017; GTF3C5; TFiiiC2-63; TFIIIC63; TFIIICepsilon
UniProt ID: (Human) Q9Y5Q8
Entrez Gene ID: (Human) 9328
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support