Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: General transcription factor 3C polypeptide 5; general transcription factor III polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; TF3C-epsilon; TFIIIC 63 kDa subunit; TFIIICepsilon; Transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD
Gene Aliases: 2700084A09Rik; CDABP0017; GTF3C5; TFiiiC2-63; TFIIIC63; TFIIICepsilon
UniProt ID: (Human) Q9Y5Q8, (Mouse) Q8R2T8
Entrez Gene ID: (Human) 9328, (Mouse) 70239
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support