Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAADERS PEDGEDEEEE EQLVLVELSG IIDSDFLSKC ENKCKVLGID TERPILQVDS CVFAGEYEDT LGTCVIFEEN VEHADTEGNN KTVLKYKCHT MKKLSMTRTL LTEKKEGEEN IGGVEWLQIK DNDFSYRPNM ICNFLHENED EEVVASAPDK SLELEEEEIQ MNDSSNLSCE QEKPMHLEIE DSGPLIDIPS ETEGSVFMET QMLP (1-213 aa encoded by BC011593)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: General transcription factor 3C polypeptide 6; general transcription factor IIIC, polypeptide 6, alpha 35kDa; TFIIIC 35 kDa subunit; Transcription factor IIIC 35 kDa subunit; transcription factor IIIC 35kDa; Transcription factor IIIC subunit 6
Gene Aliases: bA397G5.3; C6orf51; CDA020; GTF3C6; NPD020; TFIIIC35
UniProt ID: (Human) Q969F1
Entrez Gene ID: (Human) 112495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support