Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
GYS1 Monoclonal Antibody (J.18.5) detects endogenous levels of total muscle and liver glycogen synthase protein.
It is not recommended to aliquot this antibody.
Glycogen is a polysaccharide of glucose and serves as an energy storage in mammalian muscle and liver. Glycogen synthase catalyzes the rate-limiting step of glycogen biosynthesis through the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Glycogen synthase has two major isoforms in mammals: muscle isoform (GYS1) and liver isoform (GYS2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycogen [starch] synthase, liver; Glycogen [starch] synthase, muscle; Glycogen synthase 1; glycogen synthase 1 (muscle); Glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase 3, brain; glycogen synthase, liver
Gene Aliases: BC021322; GLYSN; GSY; GYS; GYS1; GYS2; Gys3; LGS; MGS
UniProt ID: (Human) P13807, (Human) P54840, (Rat) A2RRU1, (Mouse) Q9Z1E4, (Mouse) Q8VCB3
Entrez Gene ID: (Human) 2997, (Human) 2998, (Rat) 690987, (Mouse) 14936, (Mouse) 232493, (Rat) 25623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support