Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, -10, or recombinant mouse Galectin-9 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Galectin 14 binds beta-galactoside and lactose. Strong inducer of T-cell apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charcot-Leyden crystal protein 2; CLC2; Gal-14; Galectin-14; lectin, galactoside binding soluble 14; lectin, galactoside-binding, soluble, 14; MGC22235; Placental protein 13-like; placental protein 13-like protein
Gene Aliases: CLC2; LGALS14; PPL13
UniProt ID: (Human) Q8TCE9
Entrez Gene ID: (Human) 56891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support