Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bombesin; Gastrin-releasing peptide; GRP; neuromedin C; pre-progastrin releasing peptide; prepro-GRP; testicular tissue protein Li 103
Gene Aliases: BLP; BN; GRP; GRP-10; preproGRP; proGRP
UniProt ID: (Human) P07492, (Rat) P24393, (Mouse) Q8R1I2
Entrez Gene ID: (Human) 2922, (Rat) 171101, (Mouse) 225642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support