Product References
GM-CSF-activated human dendritic cells promote type 1 T follicular helper cell polarization in a CD40-dependent manner.
Journal of cell science
Korniotis S,Saichi M,Trichot C,Hoffmann C,Amblard E,Viguier A,Grondin S,Noel F,Mattoo H,Soumelis V
25-9966-42 was used in Flow cytometry/Cell sorting to report that human blood CD1c+ dendritic cells activated by GM-CSF (also known as CSF2) drive the differentiation of naive CD4+ T cells into Tfh1 cells.
Tue Nov 01 00:00:00 UTC 2022
GM-CSF-activated human dendritic cells promote type 1 T follicular helper cell polarization in a CD40-dependent manner.
Journal of cell science
Korniotis S,Saichi M,Trichot C,Hoffmann C,Amblard E,Viguier A,Grondin S,Noel F,Mattoo H,Soumelis V
25-9966-42 was used in Flow cytometry/Cell sorting to report that human blood CD1c+ dendritic cells activated by GM-CSF (also known as CSF2) drive the differentiation of naive CD4+ T cells into Tfh1 cells.
Tue Nov 01 00:00:00 UTC 2022
Airway Administration of Bacterial Lysate OM-85 Protects Mice Against Respiratory Syncytial Virus Infection.
Frontiers in immunology
Antunes KH,Cassão G,Santos LD,Borges SG,Poppe J,Gonçalves JB,Nunes EDS,Recacho GF,Sousa VB,Da Silva GS,Mansur D,Stein RT,Pasquali C,De Souza APD
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Tue May 24 00:00:00 UTC 2022
The molecular and epigenetic mechanisms of innate lymphoid cell (ILC) memory and its relevance for asthma.
The Journal of experimental medicine
Verma M,Michalec L,Sripada A,McKay J,Sirohi K,Verma D,Sheth D,Martin R,Dyjack N,Seibold MA,Knapp JR,Tu TH,O'Connor BP,Gorska MM,Alam R
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Mon Jul 05 00:00:00 UTC 2021
Histone deacetylase 3 represses cholesterol efflux during CD4+ T-cell activation.
eLife
Wilfahrt D,Philips RL,Lama J,Kizerwetter M,Shapiro MJ,McCue SA,Kennedy MM,Rajcula MJ,Zeng H,Shapiro VS
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Thu Dec 02 00:00:00 UTC 2021
Effector T cell responses unleashed by regulatory T cell ablation exacerbate oral squamous cell carcinoma.
Cell reports. Medicine
Chao JL,Korzinkin M,Zhavoronkov A,Ozerov IV,Walker MT,Higgins K,Lingen MW,Izumchenko E,Savage PA
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Tue Sep 21 00:00:00 UTC 2021
LmjF.36.3850, a novel hypothetical Leishmania major protein, contributes to the infection.
Immunology
Zutshi S,Sarode AY,Ghosh SK,Jha MK,Sudan R,Kumar S,Sadhale LP,Roy S,Saha B
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2021
Graded RhoA GTPase Expression in Treg Cells Distinguishes Tumor Immunity From Autoimmunity.
Frontiers in immunology
Kalim KW,Yang JQ,Modur V,Nguyen P,Li Y,Zheng Y,Guo F
25-9966-42 was used in Flow cytometry/Cell sorting to suggest that graded RhoA expression in Treg cells distinguishes tumor immunity from autoimmunity and that rational targeting of RhoA in Treg cells may trigger anti-tumor T cell immunity without causing autoimmune responses.
Tue Jan 11 00:00:00 UTC 2022
Graded RhoA GTPase Expression in Treg Cells Distinguishes Tumor Immunity From Autoimmunity.
Frontiers in immunology
Kalim KW,Yang JQ,Modur V,Nguyen P,Li Y,Zheng Y,Guo F
25-9966-42 was used in Flow cytometry/Cell sorting to suggest that graded RhoA expression in Treg cells distinguishes tumor immunity from autoimmunity and that rational targeting of RhoA in Treg cells may trigger anti-tumor T cell immunity without causing autoimmune responses.
Tue Jan 11 00:00:00 UTC 2022
BNT162b vaccines protect rhesus macaques from SARS-CoV-2.
Nature
Vogel AB,Kanevsky I,Che Y,Swanson KA,Muik A,Vormehr M,Kranz LM,Walzer KC,Hein S,Güler A,Loschko J,Maddur MS,Ota-Setlik A,Tompkins K,Cole J,Lui BG,Ziegenhals T,Plaschke A,Eisel D,Dany SC,Fesser S,Erbar S,Bates F,Schneider D,Jesionek B,Sänger B,Wallisch AK,Feuchter Y,Junginger H,Krumm SA,Heinen AP,Adams-Quack P,Schlereth J,Schille S,Kröner C,de la Caridad Güimil Garcia R,Hiller T,Fischer L,Sellers RS,Choudhary S,Gonzalez O,Vascotto F,Gutman MR,Fontenot JA,Hall-Ursone S,Brasky K,Griffor MC,Han S,Su AAH,Lees JA,Nedoma NL,Mashalidis EH,Sahasrabudhe PV,Tan CY,Pavliakova D,Singh G,Fontes-Garfias C,Pride M,Scully IL,Ciolino T,Obregon J,Gazi M,Carrion R,Alfson KJ,Kalina WV,Kaushal D,Shi PY,Klamp T,Rosenbaum C,Kuhn AN,Türeci Ö,Dormitzer PR,Jansen KU,Sahin U
25-9966-42 was used in flow cytometry to report the preclinical development of two vaccine candidates (BNT162b1 and BNT162b2) that contain nucleoside-modified messenger RNA that encodes immunogens derived from the spike glycoprotein (S) of SARS-CoV-2, formulated in lipid nanoparticles.
Thu Apr 01 00:00:00 UTC 2021
BNT162b vaccines protect rhesus macaques from SARS-CoV-2.
Nature
Vogel AB,Kanevsky I,Che Y,Swanson KA,Muik A,Vormehr M,Kranz LM,Walzer KC,Hein S,Güler A,Loschko J,Maddur MS,Ota-Setlik A,Tompkins K,Cole J,Lui BG,Ziegenhals T,Plaschke A,Eisel D,Dany SC,Fesser S,Erbar S,Bates F,Schneider D,Jesionek B,Sänger B,Wallisch AK,Feuchter Y,Junginger H,Krumm SA,Heinen AP,Adams-Quack P,Schlereth J,Schille S,Kröner C,de la Caridad Güimil Garcia R,Hiller T,Fischer L,Sellers RS,Choudhary S,Gonzalez O,Vascotto F,Gutman MR,Fontenot JA,Hall-Ursone S,Brasky K,Griffor MC,Han S,Su AAH,Lees JA,Nedoma NL,Mashalidis EH,Sahasrabudhe PV,Tan CY,Pavliakova D,Singh G,Fontes-Garfias C,Pride M,Scully IL,Ciolino T,Obregon J,Gazi M,Carrion R,Alfson KJ,Kalina WV,Kaushal D,Shi PY,Klamp T,Rosenbaum C,Kuhn AN,Türeci Ö,Dormitzer PR,Jansen KU,Sahin U
25-9966-42 was used in flow cytometry to report the preclinical development of two vaccine candidates (BNT162b1 and BNT162b2) that contain nucleoside-modified messenger RNA that encodes immunogens derived from the spike glycoprotein (S) of SARS-CoV-2, formulated in lipid nanoparticles.
Thu Apr 01 00:00:00 UTC 2021
BNT162b vaccines protect rhesus macaques from SARS-CoV-2.
Nature
Vogel AB,Kanevsky I,Che Y,Swanson KA,Muik A,Vormehr M,Kranz LM,Walzer KC,Hein S,Güler A,Loschko J,Maddur MS,Ota-Setlik A,Tompkins K,Cole J,Lui BG,Ziegenhals T,Plaschke A,Eisel D,Dany SC,Fesser S,Erbar S,Bates F,Schneider D,Jesionek B,Sänger B,Wallisch AK,Feuchter Y,Junginger H,Krumm SA,Heinen AP,Adams-Quack P,Schlereth J,Schille S,Kröner C,de la Caridad Güimil Garcia R,Hiller T,Fischer L,Sellers RS,Choudhary S,Gonzalez O,Vascotto F,Gutman MR,Fontenot JA,Hall-Ursone S,Brasky K,Griffor MC,Han S,Su AAH,Lees JA,Nedoma NL,Mashalidis EH,Sahasrabudhe PV,Tan CY,Pavliakova D,Singh G,Fontes-Garfias C,Pride M,Scully IL,Ciolino T,Obregon J,Gazi M,Carrion R,Alfson KJ,Kalina WV,Kaushal D,Shi PY,Klamp T,Rosenbaum C,Kuhn AN,Türeci Ö,Dormitzer PR,Jansen KU,Sahin U
25-9966-42 was used in flow cytometry to report the preclinical development of two vaccine candidates (BNT162b1 and BNT162b2) that contain nucleoside-modified messenger RNA that encodes immunogens derived from the spike glycoprotein (S) of SARS-CoV-2, formulated in lipid nanoparticles.
Thu Apr 01 00:00:00 UTC 2021
Bendamustine with total body irradiation conditioning yields tolerant T-cells while preserving T-cell-dependent graft-versus-leukemia.
Oncoimmunology
Stokes J,Hoffman EA,Molina MS,Kummet N,Simpson RJ,Zeng Y,Katsanis E
25-9966 was used in Flow cytometry/Cell sorting to demonstrate that BEN-TBI reduces graft-versus-host disease (GvHD) compared to CY-TBI independently of T regulatory cells (Tregs).
Wed Jul 14 00:00:00 UTC 2021
Bendamustine with total body irradiation conditioning yields tolerant T-cells while preserving T-cell-dependent graft-versus-leukemia.
Oncoimmunology
Stokes J,Hoffman EA,Molina MS,Kummet N,Simpson RJ,Zeng Y,Katsanis E
25-9966 was used in Flow cytometry/Cell sorting to demonstrate that BEN-TBI reduces graft-versus-host disease (GvHD) compared to CY-TBI independently of T regulatory cells (Tregs).
Wed Jul 14 00:00:00 UTC 2021
Attenuation of TCR-induced transcription by Bach2 controls regulatory T cell differentiation and homeostasis.
Nature communications
Sidwell T,Liao Y,Garnham AL,Vasanthakumar A,Gloury R,Blume J,Teh PP,Chisanga D,Thelemann C,de Labastida Rivera F,Engwerda CR,Corcoran L,Kometani K,Kurosaki T,Smyth GK,Shi W,Kallies A
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Tue Jan 14 00:00:00 UTC 2020
The obesity-induced adipokine sST2 exacerbates adipose Treg and ILC2 depletion and promotes insulin resistance.
Science advances
Zhao XY,Zhou L,Chen Z,Ji Y,Peng X,Qi L,Li S,Lin JD
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
B cells induced by Schistosoma japonicum infection display diverse regulatory phenotypes and modulate CD4+ T cell response.
Parasites & vectors
Xiao J,Guan F,Sun L,Zhang Y,Zhang X,Lu S,Liu W
25-9966-42 was used in Flow Cytometry to show that Schistosoma japonicum infection modulates the differentiation of B cell subsets that have the capability to affect the CD4+ T cell response.
Fri Mar 20 00:00:00 UTC 2020
Compartmentalized gut lymph node drainage dictates adaptive immune responses.
Nature
Esterházy D,Canesso MCC,Mesin L,Muller PA,de Castro TBR,Lockhart A,ElJalby M,Faria AMC,Mucida D
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Wed May 01 00:00:00 UTC 2019
TREM-1-dependent M1 macrophage polarization restores intestinal epithelium damaged by DSS-induced colitis by activating IL-22-producing innate lymphoid cells.
Journal of biomedical science
Yang FC,Chiu PY,Chen Y,Mak TW,Chen NJ
25-9966 was used in Flow cytometry to suggest that TREM-1 plays a crucial role in regulating IL-22 production by ILC3 through modulating M1-macrophage polarization during DSS-induced acute colitis.
Wed Jun 12 00:00:00 UTC 2019
IL-33-mediated mast cell activation promotes gastric cancer through macrophage mobilization.
Nature communications
Eissmann MF,Dijkstra C,Jarnicki A,Phesse T,Brunnberg J,Poh AR,Etemadi N,Tsantikos E,Thiem S,Huntington ND,Hibbs ML,Boussioutas A,Grimbaldeston MA,Buchert M,O'Donoghue RJJ,Masson F,Ernst M
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Fri Jun 21 00:00:00 UTC 2019
Establishment of Chronic Typhoid Infection in a Mouse Carriage Model Involves a Type 2 Immune Shift and T and B Cell Recruitment to the Gallbladder.
mBio
González JF,Kurtz J,Bauer DL,Hitt R,Fitch J,Wetzel A,La Perle K,White P,McLachlan J,Gunn JS
25-9966 was used in Flow cytometry/Cell sorting to analyze the transcriptional profile of the gallbladder in a mouse model of chronic carriage.
Tue Oct 01 00:00:00 UTC 2019
Stability and function of regulatory T cells expressing the transcription factor T-bet.
Nature
Levine AG,Mendoza A,Hemmers S,Moltedo B,Niec RE,Schizas M,Hoyos BE,Putintseva EV,Chaudhry A,Dikiy S,Fujisawa S,Chudakov DM,Treuting PM,Rudensky AY
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Thu Jun 15 00:00:00 UTC 2017
Regulatory Innate Lymphoid Cells Control Innate Intestinal Inflammation.
Cell
Wang S,Xia P,Chen Y,Qu Y,Xiong Z,Ye B,Du Y,Tian Y,Yin Z,Xu Z,Fan Z
25-9966 was used in Flow cytometry/Cell sorting to identify a regulatory subpopulation of innate lymphoid cells that exists in the gut, and harbour a unique gene identity.
Thu Sep 21 00:00:00 UTC 2017
ILC2s regulate adaptive Th2 cell functions via PD-L1 checkpoint control.
The Journal of experimental medicine
Schwartz C,Khan AR,Floudas A,Saunders SP,Hams E,Rodewald HR,McKenzie ANJ,Fallon PG
25-9966 was used in Flow cytometry/Cell sorting to study how group 2 innate lymphoid cells regulate adaptive Th2 cell functions.
Mon Sep 04 00:00:00 UTC 2017
Characterisation of innate lymphoid cell populations at different sites in mice with defective T cell immunity.
Wellcome open research
Dutton EE,Camelo A,Sleeman M,Herbst R,Carlesso G,Belz GT,Withers DR
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Sat Jan 09 00:00:00 UTC 2021
Antibiotic-induced perturbations in microbial diversity during post-natal development alters amyloid pathology in an aged APPSWE/PS1ΔE9 murine model of Alzheimer's disease.
Scientific reports
Minter MR,Hinterleitner R,Meisel M,Zhang C,Leone V,Zhang X,Oyler-Castrillo P,Zhang X,Musch MW,Shen X,Jabri B,Chang EB,Tanzi RE,Sisodia SS
25-9966 was used in Flow cytometry/Cell sorting to investigate how ABX-induced microbial diversity perturbations during post-natal stages of development coincide with altered host immunity mechanisms and amyloidosis in a murine model of AD.
Tue Sep 05 00:00:00 UTC 2017
Intranasal Administration of Lentiviral miR-135a Regulates Mast Cell and Allergen-Induced Inflammation by Targeting GATA-3.
PloS one
Deng YQ,Yang YQ,Wang SB,Li F,Liu MZ,Hua QQ,Tao ZZ
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Tue May 31 00:00:00 UTC 2016
Impaired proteasome function activates GATA3 in T cells and upregulates CTLA-4: relevance for Sézary syndrome.
The Journal of investigative dermatology
Gibson HM,Mishra A,Chan DV,Hake TS,Porcu P,Wong HK
Published figure using Gata-3 monoclonal antibody (Product # 25-9966-42) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2013
Role of CD44 in the differentiation of Th1 and Th2 cells: CD44-deficiency enhances the development of Th2 effectors in response to sheep RBC and chicken ovalbumin.
Journal of immunology (Baltimore, Md. : 1950)
Guan H,Nagarkatti PS,Nagarkatti M
Wed Jul 01 00:00:00 UTC 2009