Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAQHGMDD DGTGQKQIWR IEGSNKVPVD PATYGQFYGG DSYIILYNYR HGGRQGQIIY NWQGAQSTQD EVAASAILTA QLDEELGGTP VQSRVVQGKE PAHLMSLFGG KPMIIYKGGT SREGGQTAPA STRLFQVRAN SAGATRAVEV LPKAGALNSN DAFVLKTPSA AYLWVGTGAS EAEKTGAQEL LRVLRAQPVQ VAEGSEPDGF WEALGGKAAY RTSPRLKDKK MDAHPPRLFA CSNKIGRFVI EEVPGELMQE DLATDDVMLL DTWDQVFVWV GKDSQEEEKT EALTSAKRYI ETDPANRDRR TPITVVKQGF EPPSFVGWFL GWDDDYWSVD PLDRAMAELA A (433-782 aa encoded by BC026033)
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; Gelsolin; RP11-477J21.1
Gene Aliases: ADF; AGEL; GSN
UniProt ID: (Human) P06396
Entrez Gene ID: (Human) 2934
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support