Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: ECPIAQESLE SQEQRARAAL RERYLRSLLA MVGHQVSFTL HEGVRVAAHF GATDLDVANF YVSQLQTPIG VQAEALLRCS DIISYTFKP
The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear "gems" (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gem (nuclear organelle) associated protein 7; Gem-associated protein 7; gemin 7; Gemin-7; KIAA2024; SIP3; ZNF650
Gene Aliases: GEMIN7; SIP3
UniProt ID: (Human) Q9H840
Entrez Gene ID: (Human) 79760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support