Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN2A; GluRepsilon1; Glutamate [NMDA] receptor subunit epsilon-1; glutamate receptor; Glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, N-methyl D-aspartate 2A; hNR2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NMDA Receptor 2A; NMDAR 2A; NMDAR2A; OTTHUMP00000160135; OTTHUMP00000174531
Gene Aliases: EPND; FESD; GluN2A; GRIN2A; LKS; NMDAR2A; NR2A
UniProt ID: (Human) Q12879, (Mouse) P35436, (Rat) Q63728
Entrez Gene ID: (Human) 2903, (Rabbit) 100337889, (Mouse) 14811, (Rat) 24409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support