Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
The NMDA Receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimers, epilepsy and ischemic neuronal cell death. The NMDA receptor is also one of the principal molecular targets for alcohol in the CNS. The NMDAR is also potentiated by protein phosphorylation. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of th NMDAR2 (NR2 A-D) subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN2C; GluRepsilon3; Glutamate [NMDA] receptor subunit epsilon-3; glutamate receptor; Glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, N-methyl D-aspartate 2C; glutamate receptor, ionotropic, NMDA2C; N-methyl D-aspartate receptor subtype 2C; N-methyl-D-aspartate receptor subunit 2C; NMDA glutamate receptor; NMDAR2C; NR2
Gene Aliases: GluN2C; GRIN2C; NMDAR2C; NR2C
UniProt ID: (Human) Q14957, (Mouse) Q01098, (Rat) Q00961
Entrez Gene ID: (Human) 2905, (Mouse) 14813, (Rat) 24411
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support