Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GKRP; Glucokinase regulator; Glucokinase regulatory protein; hexokinase 4
Gene Aliases: FGQTL5; GCKR; GKRP; GLRE
UniProt ID: (Human) Q14397, (Rat) Q07071, (Mouse) Q91X44
Entrez Gene ID: (Human) 2646, (Rat) 25658, (Mouse) 231103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support