Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; GLU2B; Glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; Glucosidase II subunit beta; hepatocystin; PKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate, 80 Kda protein
Gene Aliases: 80K-H; AGE-R2; G19P1; GIIB; PCLD; PKCSH; PLD1; PRKCSH; VASAP-60
UniProt ID: (Human) P14314, (Mouse) O08795
Entrez Gene ID: (Human) 5589, (Mouse) 19089, (Rat) 300445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support