Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. 자세히 알아보기
FIGURE: 1 / 7
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASTP; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; ATP:glycerol 3-phosphotransferase; EC 2.7.1.30; GK 3; GK3P; GKl; GKP3; GKTB; Glycerokinase; Glycerokinase 3; Glycerol kinase; Glycerolkinase
Gene Aliases: ASTP; D930012N15Rik; GK; GK1; GKD; Gyk
UniProt ID: (Human) P32189, (Rat) Q63060, (Mouse) Q64516
Entrez Gene ID: (Human) 2710, (Rat) 79223, (Mouse) 14933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support