Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
The GLRA3 gene encodes the alpha-3 subunit of the neuronal glycine receptor, a ligand-gated chloride channel composed of ligand-binding alpha and structural beta polypeptides.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: glycine receptor; Glycine receptor subunit alpha-3; glycine receptor, alpha 3; glycine receptor, alpha-3 polypeptide; ligand gated ion channel
基因别名: GLRA3
UniProt ID: (Human) O75311
Entrez Gene ID: (Human) 8001