Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 15
Recommended positive controls: A431, K562, MCF-7, AsPC-1, MDA-MB-231, MDA-MB-231 (24 µg/mL Tunicamycin for 16hr), U87-MG, U87-MG membrane extracts, U87-MG conditioned medium, Mouse brain, mouse fetal brain, A431 (500 units PNGase F for 4 hr), A431 (24 µg/mL Tunicamycin treatment for 16hr), A431 (4 mM Benzyl glucosinolate treatment for 32hr), A431(24 µg/mL Tunicamycin treatment for 16hr and 4 mM Benzyl glucosinolate treatment for 32hr).
Predicted reactivity: Mouse (89%), Rat (88%), Rhesus Monkey (97%), Bovine (89%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glypican proteoglycan 1; Glypican-1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12
Gene Aliases: AI462976; glypican; GPC1; HSPG M12; HSPGM12
UniProt ID: (Human) P35052, (Mouse) Q9QZF2, (Rat) P35053
Entrez Gene ID: (Human) 2817, (Mouse) 14733, (Rat) 58920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support